Learn More
Rotavirus Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand:  Invitrogen PA185845
Product Code. 11513721
  • 498.00 € / 1mL
Explore more special offers
Description
Description
PA1-85845 detects human rotavirus serotypes I, II, III, IV, and VIII. Sheep anti Rotavirus antibody recognizes human rotavirus serotypes I, II, III, IV and VIII.
Rotavirus is a double-stranded RNA virus in the family Reoviridae. Five species of rotavirus (A-E) cause disease in animals with species A being the most common in human. Within rotavirus A there are different strains, called serotypes, based on the dual classification system of two proteins on the surface of the virus. Six viral proteins (VPs), or structural proteins, form the virion and six non-structural proteins (NSPs) are expressed in infected cells. Rotavirus is the most common cause of severe diarrhoea among infants and young children. Rotavirus gastroenteritis is a mild to severe disease characterised by vomiting, watery diarrhoea, and low-grade fever, but still leads to hundreds of thousands of deaths each year in developing countries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotavirus
Polyclonal
Unconjugated
Reoviridae
Five serotypes of group A Rotavirus strains Wa (serotype I), DS-1 (serotype II), Ito (III), HOCHI (IV) and 69M (VIII)
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunocytochemistry
5 mg/mL
PBS with 0.09% sodium azide
Sheep
Protein G
RUO
Virus