missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotenone, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Mitochondrial electron transport chain inhibitor (IC50 = 1.7 - 2.2 μM at complex I). Inhibits NADH oxidation by cardiac sarcoplasmic reticulum (IC50 = 3.4 nM). Commonly used pesticide and induces Parkinsonism in animal models. May also inhibit autophagy induction; blocks lysosomal degradation of autophagic vacuoles. Cell-permeable and brain penetrant.
Specifications
Specifications
CAS | 83-79-4 |
Molecular Formula | C23H22O6 |
Quantity | 50 mg |
Synonym | rotenone, dactinol, paraderil, --rotenone, tubatoxin, barbasco, --cis-rotenone, derrin, derris, cube root |
InChI Key | JUVIOZPCNVVQFO-HBGVWJBISA-N |
SMILES | CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC |
Molecular Weight (g/mol) | 394.423 |
PubChem CID | 6758 |
ChEBI | CHEBI:28201 |
Formula Weight | 394.42 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction