missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries SI™ Roto-Shake Genie Rotato Rocker 230V, without plug

Heavy duty Multi Purpose Rotator that turns into a rocker by pressing a button.

Brand:  Scientific Industries SI™ SI-1101

Product Code. 12613125

  • 1305.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rotator turns into rocker by pressing a button
  • Without plug
Specifications

Specifications

4.5 kg
230 V
0.5 A
Rotator Rocker
CE
12 in.
508 x 252 x 267 mm
267 mm
508 mm
9.9 in
200 mm
14.52 kg
11.8 kg
Provide rotating, rocking, and orbital shaking in one compact, durable and user-friendly unit
0.95
5 to 35 rpm
230 V
12 x 10 to 13 mm Tubes, 6 x 15 to 17 mm Tubes, 3 x 28 to 30 mm Tubes
10 lb.
50 Hz
305 mm
10.5 in
20 in
340 mm
252 mm
32 lb
26 lb
Roto-shake Genie without Plug
Clip plates, Metal rocking tray, magnetic strips, non-skid tray
2 year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.