missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Rotor for Centrifuge 5420
Eppendorf Rotors for Centrifuge 5420, precision for diverse applications. Choose from various capacities and speeds to tailor your centrifuge setup. Accessorize with adaptors and buckets, ensuring optimal sample processing in molecular biology and beyond.
Brand: Eppendorf™ 5495404007
530.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight options available
Specifications
F-32x0.2-PCR - Includes rotor lid | |
Rotor | |
18,210 x g | |
Centrifuge 5421 |
Anodized Aluminum | |
4 x PCR-strips or 32 x PCR Tubes (0.2 mL) | |
14,800 rpm | |
Rotor F-32x0.2-PCR for Centrifuge 5420 |