missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R

Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.

Brand:  Eppendorf™ 5895120008

3211.00 EUR valid until 2024-12-31
Use promo code "21988" to get your promotional price.


 View more versions of this product

Product Code. 15758369

This item is not returnable. View return policy

Description

Description

  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S-4x750, Aerosol-tight - Includes round buckets
4 x 750 mL
4,700 rpm
Round buckets
Rotor S-4x750, incl. 4 × 750 mL round buckets
4,800 x g
Centrifuges 5910 Ri and 5920 R
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers