Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
1550.00 € - 4905.00 €
Specifications
Type Rotor
For Use With (Equipment) Centrifuges 5910 Ri and 5920 R
View More Specs
Products 8
Product Code Brand Description Capacity Max. RCF Max. Speed Price Quantity & Availability  
Product Code Brand Description Capacity Max. RCF Max. Speed Price Quantity & Availability  
15718379
View accessory products View Documents
Eppendorf™
5895130003
FA-20x5, Aerosol-tight - Includes Eppendorf QuickLock™ rotor lid 20 × 5 mL 20,913 x g 13,100 rpm
1580.00 €
Each
15758369
View accessory products View Documents
Eppendorf™
5895120008
S-4x750, Aerosol-tight - Includes round buckets 4 x 750 mL 4,800 x g 4,700 rpm
3380.00 €
Each
15768369
View Documents
Eppendorf™
5895121004
S-4x750, Aerosol-tight - without buckets 4 x 750 mL 4,800 x g 4,700 rpm
1550.00 €
Each
15633797
View accessory products View Documents
Eppendorf™
5895135005
FA-48x2, Aerosol-tight - Includes Eppendorf QuickLock™ rotor lid 48 x 2 mL 21,194 x g 13,700 rpm
1616.44 €
Each
15674614
View accessory products View Documents
Eppendorf™
5895150004
FA-6x50, Aerosol-tight - Includes rotor lid 6 × 50 mL 20,130 x g 13,500 rpm
2575.00 €
Each
15863212
View accessory products View Documents
Eppendorf™
5895200001
S-4xUniversal - Includes universal buckets 4 to 750 mL 4,347 x g 4500 rpm
4010.00 €
Each
15873212
View Documents
Eppendorf™
5895201008
S-4xUniversal - without buckets 4 to 750 mL 4,347 x g 4500 rpm
1790.00 €
Each
16202411
View accessory products View Documents
Eppendorf™
5895175007
FA-6x250, Aerosol-tight - Includes Eppendorf QuickLock™ rotor lid 6 x 250 mL 15,054 x g 10,100 rpm
4905.00 €
Each
Description
Description
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
  • Aerosol-tight options available.
Specifications
Specifications
Rotor
Centrifuges 5910 Ri and 5920 R