missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Round Basin

Round Basin for food contact. Highly durable and chemical resistant.

11.15 € - 19.55 €

Specifications

Type Storage Tub
Material LDPE
Color White
Shape Circular
Certifications/Compliance Food Contact Certified
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
13419898
View Documents
Gilac™
G606521
4.5 L
11.15 €
Each
In Stock
to see stock.
Add to basket
 
13429898
View Documents
Gilac™
G606721
7.5 L
15.35 €
Each
In Stock
to see stock.
Edge
Add to basket
 
13439898
View Documents
Gilac™
G606821
9 L
19.55 €
Each
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
 
Description

Description

  • Basin fit for food contact
  • Highly durable and chemical resistant
  • Easy to clean
  • Nesting with no suction effect
  • Moulded rim makes it easy to grasp
Specifications

Specifications

Storage Tub
White
Food Contact Certified
Nestable
-15°C to +60°C
LDPE
Circular
Food Contact
5°F to 140°F
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilac™ Round Basin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.