Learn More
Lenz Laborglasintrumente Round Bottom Flasks, Narrow Neck
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07030073
 View more versions of this product
Product Code. 10214441
  • 644.00 € / Pack of 10
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN ISO 1773 and in addition to standard
  • With beaded rim
  • Material: DURAN™
Specifications
Specifications
42 mm
260 mm
10/Pk.
2 L
Beaded rim