Learn More
Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation
Ideal for routine laboratory analysis and procedures. Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation are manufactured from durable DURAN™ borosilicate glass.
45.00 € - 52.00 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10396412
|
Lenz Laborglasinstrumente™
05700007 |
Neck Size: 14/23; Without Stopper |
52.00 €
Pack of 10 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||||
10767571
|
Lenz Laborglasinstrumente™
05700107 |
Neck Size: 14/23; Material: Polyethylene; Without Stopper |
45.00 €
Pack of 10 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||||
Description
- With round bottom and NS socket
- Available without stopper, with PE stopper, or with hexagonal hollow glass stopper
- Corrosion-free and resistant to thermal shock
Specifications
Glass Tube |