Learn More
Thermo Scientific™ Round Bucket for Thermo Scientific™ Sorvall™ Stratos Rotor
For use with Four-Place Swinging Bucket Rotor 75-003-047
Brand:  Thermo Scientific™ 75008172
Product Code. 10488732
  • 404.01 € / Each
Explore more special offers
Description
Description
Order Info
Order correct quantity to equip rotor. Aerosol-tight lids (75008173) sold separately. Optional adapters accept assorted tube styles and volumes from 1.5mL to 100mL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Swing out rotor for Labofuge™ 400 and 400R centrifuge
18 mL
1.5 to 100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products