Learn More
Fisherbrand™ Cylindrical Stir Bars with Removable Pivot Ring
Combining top-notch and uncompromising quality with an affordable price. Fisherbrand™ Round Magnetic Stir Bars are reliable and dependable.
Brand:  Fisherbrand™ F37111-5002
Product Code. 15535604
  • 4.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Teflon™ resin-coated
  • Removable pivot ring
Specifications
Specifications
Autoclavable
White
Removable pivot ring
8 mm
Alnico Magnet
FDA, ISO 9001:2008
0.31in.
2in.
50.8 mm
Cylinder