Learn More
Thermo Scientific™ Round Test Tube Cells for Thermo Scientific Spectronic™ 20+ and 20D+ Spectrophotometers
Brand:  Thermo Scientific™ 331782-000
 View more versions of this product
Product Code. 11893774
  • 545.00 € / Pack of 12
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optical class
  • Cat. No. 07-144-15 1/2 in. diameter test tube cell replacements for originals supplied with instrument Cat. No. 07-144-40 3/4 in. diameter test tube cells require 07-144-20 adapter and 07-144-24 light shield (sold separately)
  • Cat. No. 07-144-38 test tube cell is 1 in. diameter and requires 07-144-22 adapter and 07-144-24 light shield (sold separately)
  • Packaged in box of 12
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Test Tube
25.4 mm
Thermo Scientific Spectronic™ 20 and 20D spectrophotometers
Optical Glass
150 mm