missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Routine DNA Ladder

Ready-to-use DNA size standard is room-temperature stable for up to two years

164.07 € - 219.00 €

Specifications

Color Blue
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Quantity 1000 μL
View More Specs

Products 2
Product Code Brand For Use With (Application) Product Type No. of Bands Price Quantity & Availability  
Product Code Brand For Use With (Application) Product Type No. of Bands Price Quantity & Availability  
10284633
View Documents
Fisher BioReagents™
BP2581-200
PCR size confirmation DNA Molecular Weight Marker/Ladder 11 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10450464
View Documents
Fisher BioReagents™
BP2582-200
General purpose, wide DNA size range DNA Molecular Weight Marker/Ladder 16 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Includes a landmark doublet at 1400 and 1550 bp

Specifications

Specifications

Blue
DNA Molecular Weight Marker/Ladder
Liquid
1000 μL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Low Scale DNA Ladder, 100bp

SDS
Documents

Documents

Product Certifications