missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL26L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16487-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that shares high sequence similarity with ribosomal protein L26. Alternative splicing results in multiple transcript variants encoding the same protein.Specifications
RPL26L1 | |
Polyclonal | |
Unconjugated | |
RPL26L1 | |
ribosomal protein L26 like 1, RPL26L1, RPL26P1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
307636, 51121, 668405 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UNX3 | |
RPL26L1, Rpl26-ps3 | |
RPL26L1 Fusion Protein Ag9609 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction