Learn More
Corning™ RPMI 1640, 1X
Glutagro™ supplement provides greatly improved stability over the L-glutamine media supplement
180.45 €
Product Code | Brand | Product Type | pH | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | pH | Quantity | Price | Quantity & Availability | |||
15363561
|
Corning™
17-105-CV |
RPMI 1640 w/o L-glut and ph.red | 7 to 7.4 | 6/Pk. |
180.45 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol. Cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.
- Can be used for suspension cultures or monolayer human leukemic cells
- This formulation is supplemented with glutaGRO™ in place of L-glutamine for improved stability
Specifications
2° to 8°C |
Safety and Handling
missing translation for 'shelfLife' : 18 Months