Learn More
MP Biomedicals™ RPMI 1640 Medium
General purpose enriched media with extensive applications for a vast range of mammalian cells including human myeloma, mouse hybridoma, human leukocytes, and B and T lymphocytes
18.60 € - 30.25 €
Product Code | Brand | pH | Content And Storage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH | Content And Storage | Price | Quantity & Availability | ||||
11400385
|
MP Biomedicals™
091260254 |
6.9 to 7.2 | 15° to 30°C |
30.25 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11410385
|
MP Biomedicals™
091260349 |
6.9 to 7.4 | 4°C | N/A | |||||
Description
Specifications
RPMI 1640 Medium | |
Sterile |