Learn More
RPS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11990-1-AP-150UL
 View more versions of this product
Product Code. 16889423
  • 410.00 € / 150µL
Explore more special offers
Description
Description
WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.
Specifications
Specifications
RPS3
Polyclonal
Unconjugated
RPS3
40S ribosomal protein S3, ribosomal protein S3, RPS3
Rabbit
Antigen Affinity Chromatography
RUO
27050, 6188
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23396, P62908
Rps3
RPS3 Fusion Protein Ag3918
150 μL
Primary
Human, Mouse