missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11990-1-AP-20UL
This item is not returnable.
View return policy
Description
WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.Specifications
RPS3 | |
Polyclonal | |
Unconjugated | |
RPS3 | |
40S ribosomal protein S3, ribosomal protein S3, RPS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27050, 6188 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23396, P62908 | |
Rps3 | |
RPS3 Fusion Protein Ag3918 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |