Learn More
Invitrogen™ RRAGB Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA545812
Product Code. 15596952
  • 520.00 € / 100µL
Description
Peptide sequence: IDIFTSNTYV MVVMSDPSIP SAATLINIRN ARKHFEKLER VDGPKQCLLM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 93%.
Ras-homologous GTPases constitute a large family of signal transducers that alternate between an activated, GTP-binding state and an inactivated, GDP-binding state. These proteins represent cellular switches that are operated by GTP-exchange factors and factors that stimulate their intrinsic GTPase activity. All GTPases of the Ras superfamily have in common the presence of six conserved motifs involved in GTP/GDP binding, three of which are phosphate-/magnesium-binding sites (PM1-PM3) and three of which are guanine nucleotide-binding sites (G1-G3). Transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RRAGB
Polyclonal
Unconjugated
RRAGB
bA465E19.1; GTP-binding protein ragB; LOC100724885; Rag B; RAGB; Ras related GTP binding B; Ras-related GTP binding B; ras-related GTP-binding protein A; ras-related GTP-binding protein B; ras-related GTP-binding protein B; RGD:619805}; RRAGB; rragB {ECO:0000312
Rabbit
Affinity Chromatography
RUO
10325
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q5VZM2
RRAGB
Synthetic peptide directed towards the C-terminal of human RRAGB (aa 324-373).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt