missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22015-1-AP-150UL
This item is not returnable.
View return policy
Description
Analysis of cDNA clones indicates that ribosome binding protein 1 may exist in different forms due to removal of tandem repeats, or partial intraexonic splicing of RRBP1. The form presented here is lacking the canine p180 ribosome-binding domain, NQGKKAEGAQ, which is tandemly repeated close to the N-terminus in other forms that haven′t been fully characterized. RRBP1 has been excluded as a candidate gene in the cause of Alagille syndrome. Alternate splicing results in multiple transcript variants.Specifications
RRBP1 | |
Polyclonal | |
Unconjugated | |
RRBP1 | |
ES/130, ES/130 related protein, ES130, hES, KIAA1398, Ribosome binding protein 1, Ribosome receptor protein, RRBP1, RRp | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6238, 81910 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99PL5, Q9P2E9 | |
RRBP1 | |
RRBP1 Fusion Protein Ag16648 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |