Learn More
RRM1 Mouse anti-Human, CoraLite 488, Clone: 5H6F3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60073-100UL
This item is not returnable.
View return policy
Description
The RRM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RRM1. This antibody recognizes human antigen. The RRM1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
RRM1 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
5H6F3 | |
CoraLite 488 | |
100 μL | |
6240 | |
IgG2b |