missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11661-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the encoded protein (M2) is regulated in a cell-cycle dependent fashion.Specifications
RRM2 | |
Polyclonal | |
Unconjugated | |
RRM2 | |
R2, RR2, RR2M, RRM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20135, 362720, 6241 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11157, P31350, Q4KLN6 | |
Rrm2 | |
RRM2 Fusion Protein Ag2203 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |