missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15329-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in ribosome biogenesis. .Specifications
RRS1 | |
Polyclonal | |
Unconjugated | |
RRS1 | |
KIAA0112, RRR, RRS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23212, 297784, 59014 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1A5P2, Q15050, Q9CYH6 | |
Rrs1 | |
RRS1 Fusion Protein Ag7030 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction