missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RS 39604 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A potent and selective 5-HT4 antagonist, with a pKi of 9.1 at 5-HT4 receptors in guinea pig striatal membranes and greater than 1000-fold selectivity over 5-HT1A, 2C, 3 and D1, D2, M1, M2, AT1, B1 and α1C receptors. The ketone group gives RS 39604 a relatively long half life; it is also orally active and so suitable for in vivo studies.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 590.56 |
Percent Purity | >98% |
Chemical Name or Material | RS 39604 hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'