Learn More
BINDER™ RS 422 Interface Extension Cable
For Use With: Multi Management Software APT-COMTM
Brand:  BINDER™ 5023-0117
Product Code. 15681556
  • 680.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with incubators and chambers
  • 50 m length
Specifications
Specifications
RS 422 interface, accessories - RS 422 extension cable (50 m) between interface converter and unit or RS 232 / RS 422 plug distributor
Multi Management Software APT-COM™
Cable
50 m