missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (RS)-(Tetrazol-5-yl)glycine

Highly potent NMDA agonist

289.00 € - 1090.00 €

Specifications

CAS 138199-51-6
Assay Percent Range >98%
Solubility Soluble to 50 mM in water
Target NMDA Receptor Agonists
Molecular Formula C3H5N5O2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18095332
View Documents Promotion Details
Tocris Bioscience™
0312/50
50 mg
1090.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
18054662
View Documents Promotion Details
Tocris Bioscience™
0312/10
10 mg
289.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent NMDA receptor agonist, approximately 20 times more active than NMDA.
Specifications

Specifications

138199-51-6
Soluble to 50 mM in water
C3H5N5O2
98%
>98%
NMDA Receptor Agonists
Observed MW: 143.1
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature

SDS
Documents

Documents

Product Certifications

For Research Use Only