Learn More
Bellingham Stanley™ Rubber Gland Seals for Metal End-Cap Polarimeter Tubes
Rubber Gland Washers for Metal End Cap Tubes & Fitting Tool are spare parts for Bellingham Stanley™ polarimeter tubes.
Brand:  Bellingham Stanley™ 35-21
Product Code. 16267350
  • 44.40 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rubber seal washers for metal-ended glass polarimeter tubes
  • Accessory for Bellingham Stanley polarimeters
  • Supplied with installation tool
  • Pack of 12
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gland Seal Washer
Rubber
Metal End-Cap Polarimeter Tube
15.5 mm