missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progressive Safety™ Rubber Overshoes
33.15 €
Specifications
Color | Black |
---|---|
For Use With (Application) | Convert ordinary shoes into anti-slip SRC rated footwear |
Sole Features | Grip |