missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Rubbermaid Flat Shelf Cart

Ideal cart for general factory, health care, retail, food service, or office use. Cole-Parmer™ Rubbermaid Flat Shelf Cart features large, non-marking casters that glide easily on hard surfaces.

Brand:  Cole-Parmer™ 09348-40

Product Code. 11800323

  • 569.00 € / Each
Estimated Shipment: 01-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Foam plastic construction for strength, quiet operation, and easy cleaning
  • Easily slide heavy items on and off flat top shelf
  • Large, smooth-gliding casters won't mar hard surfaces
Specifications

Specifications

Flat-shelf Cart
112 x 65 x 85 cm
182 kg
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Rubbermaid Flat Shelf Cart > Dimensions: 112 x 65 x 85cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.