missing translation for 'onlineSavingsMsg'
Learn More

RuBi GABA trimethylphosphine, Tocris Bioscience™

Product Code. 30001537
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Caged GABA; inhibits neural activity

Modified version of RuBi-GABA (Cat. No. 3400). Suppresses cortical local field potential (LFP) activity in anesthetized mice after activation with a blue laser pulse; inhibits neural activity.

Specifications

Molecular Formula C27H33ClN5O2PRu
Quantity 10 mg
Synonym (bis(2, 2'-Bipyridine-N, N')trimethylphosphine)-4-aminobutyric acid ruthenium chloride complex
Formula Weight 627.09
Percent Purity >98%
Chemical Name or Material RuBi GABA trimethylphosphine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'