Learn More
Tocris Bioscience™ RuBi-Glutamate
Caged glutamate; excited by visible wavelengths
Brand:  Tocris Bioscience™ 3574/10
246.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18014462
  • 314.00 € / 10mg
Explore more special offers
Description
Description
RuBi-Glutamate is a ruthenium-bipyridine-trimethylphosphine caged glutamate that can be excited by visible wavelengths and releases glutamate after one- or two-photon excitation. Has high quantum efficiency and can be used at low concentrations, partly avoiding blockade of GABAergic transmission seen with other caged glutamate compounds. Displays high spatial resolution and generates excitatory responses in individual dendritic spines with physiological kinetics.
MNI-caged-L-glutamate (Cat. No. 1490) also available.
Specifications
Specifications
(bis(2,2 '-Bipyridine-N,N')trimethylphosphine)--1-aminopropane-1,3-dicarboxylic acid ruthenium(2+) complex sodium hexafluorophosphate salt
-20°C
RuBi-Glutamate
RuBi-Glutamate
2417096-44-5
10 mg
M.W. (Observed): 970.54