Learn More
Rubidium carbonate, 99.8% (metals basis), Thermo Scientific Chemicals
81.70 € - 361.00 €
Chemical Identifiers
CAS | 584-09-8 |
---|---|
Molecular Formula | CO3Rb2 |
Molecular Weight (g/mol) | 230.94 |
MDL Number | MFCD00011188 |
InChI Key | WPFGFHJALYCVMO-UHFFFAOYSA-L |
Synonym | rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate |
PubChem CID | 11431 |
IUPAC Name | rubidium(1+);carbonate |
SMILES | [Rb+].[Rb+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11320719
|
Thermo Scientific Alfa Aesar
010562.09 |
10 g |
81.70 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330719
|
Thermo Scientific Alfa Aesar
010562.18 |
50 g | |||||||
Description
Rubidium carbonate is used in glass-making by enhancing stability and durability as well as reducing its conductivity. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas. Rubidium carbonate glass has been extensively tested for use in anti-collision devices for motor vehicles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
584-09-8 | |
230.94 | |
WPFGFHJALYCVMO-UHFFFAOYSA-L | |
11431 | |
[Rb+].[Rb+].[O-]C([O-])=O |
CO3Rb2 | |
MFCD00011188 | |
rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate | |
rubidium(1+);carbonate |
Specifications
837°C | |
White | |
CO3Rb2 | |
MFCD00011188 | |
Soluble in water. | |
[Rb+].[Rb+].[O-]C([O-])=O | |
230.94 | |
230.95 | |
Odorless | |
(metals basis) |
584-09-8 | |
Powder | |
Rb2CO3 | |
rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate | |
WPFGFHJALYCVMO-UHFFFAOYSA-L | |
rubidium(1+);carbonate | |
11431 | |
99.8% | |
Very Hygroscopic | |
Rubidium carbonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-530-9
missing translation for 'rtecsNumber' : FG0650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only