missing translation for 'onlineSavingsMsg'
Learn More
Rubidium carbonate, 99%, Thermo Scientific Chemicals Product Code.: 15453047

Rubidium carbonate, 99%, Thermo Scientific Chemicals

Product Code. 15453047
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15453047

Brand: Thermo Scientific Alfa Aesar A12240.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.

Solubility
Soluble in water4500 g/L @20°C.

Notes
Hygroscopic. Store under dry inert gas. Store away from bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-09-8
Molecular Formula CO3Rb2
Molecular Weight (g/mol) 230.94
MDL Number MFCD00011188
InChI Key WPFGFHJALYCVMO-UHFFFAOYSA-L
Synonym rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate
PubChem CID 11431
SMILES [Rb+].[Rb+].[O-]C([O-])=O

Specifications

Melting Point 837°C
Quantity 100 g
Linear Formula Rb2CO3
Solubility Information Soluble in water4500g/L @20°C.
IUPAC Name rubidium(1+);carbonate
Formula Weight 230.95
Percent Purity 99%
Odor Odorless
Sensitivity Very Hygroscopic
Chemical Name or Material Rubidium carbonate
compliance-icons
Product Identifier
  • Rubidium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Carbonic acid, dirubidium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium carbonate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.