Learn More
Rubidium carbonate, 99%, Thermo Scientific Chemicals
43.70 € - 517.58 €
Chemical Identifiers
CAS | 584-09-8 |
---|---|
Molecular Formula | CO3Rb2 |
Molecular Weight (g/mol) | 230.94 |
MDL Number | MFCD00011188 |
InChI Key | WPFGFHJALYCVMO-UHFFFAOYSA-L |
Synonym | rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate |
PubChem CID | 11431 |
SMILES | [Rb+].[Rb+].[O-]C([O-])=O |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
15433047
|
Thermo Scientific Alfa Aesar
A12240.06 |
5 g | N/A | ||||||
15443047
|
Thermo Scientific Alfa Aesar
A12240.14 |
25 g | N/A | ||||||
15453047
|
Thermo Scientific Alfa Aesar
A12240.22 |
100 g |
517.58 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRubidium carbonate is used as a raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of special glass, for the manufacturing of high energy density micro cells and crystal scintillation counters. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas.
Solubility
Soluble in water4500 g/L @20°C.
Notes
Hygroscopic. Store under dry inert gas. Store away from bases, water/ moisture.
Identifiants chimiques
584-09-8 | |
230.94 | |
WPFGFHJALYCVMO-UHFFFAOYSA-L | |
11431 |
CO3Rb2 | |
MFCD00011188 | |
rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate | |
[Rb+].[Rb+].[O-]C([O-])=O |
Spécification
837°C | |
CO3Rb2 | |
MFCD00011188 | |
Soluble in water4500g/L @20°C. | |
[Rb+].[Rb+].[O-]C([O-])=O | |
230.94 | |
230.95 | |
Odorless | |
Rubidium carbonate |
584-09-8 | |
Rb2CO3 | |
rubidium carbonate, dirubidium carbonate, carbonic acid dirubidium salt, carbonic acid, dirubidium salt, dirubidium monocarbonate, rubidium 1+ carbonate, dirubidium 1+ ion carbonate, potassium carbonate anhydrous, rubidiumcarbonate | |
WPFGFHJALYCVMO-UHFFFAOYSA-L | |
dirubidium(1+) carbonate | |
11431 | |
99% | |
Very Hygroscopic |
Sécurité et traitement
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-530-9
missing translation for 'rtecsNumber' : FG0650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only