missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, 99.8% (metals basis), Thermo Scientific Chemicals

110.55 € - 427.13 €

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+);chloride
SMILES [Cl-].[Rb+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11340719
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010563.09
10 g
110.55 €
10g
Please to purchase this item. Need a web account? Register with us today!
11350719
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010563.18
50 g
427.13 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rubidium chloride is the mostly used rubidium compound, and finds use in various fields ranging from electrochemistry to molecular biology. It is an efficient non-invasive biomarker; increases dopamine and norepinephrine levels, and useful for anergic and apathetic depressives. Pharmaceutical compositions have been used as antidepressants in Europe. It is employed in biochemistry to induce cells to take up DNA. This compound increases the competence of the cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-11-9
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
rubidium(1+);chloride
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
CHEBI:78672
[Cl-].[Rb+]

Specifications

1390°C
7791-11-9
99.8% (metals basis)
RbCl
14,8284
Soluble in water. Sparingly soluble in methanol.
[Cl-].[Rb+]
120.92
CHEBI:78672
99.8%
Hygroscopic
(metals basis)
718°C
Crystalline
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+);chloride
62683
120.92
1.493
2.800
Rubidium chloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-240-9

missing translation for 'rtecsNumber' : VL8575000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only