Learn More
Rubidium chloride, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Description
Rubidium chloride is the mostly used rubidium compound, and finds its applications in various fields ranging from electrochemistry to molecular biology. It is used as a non-invasive biomarker, anergic and apathetic depressives in the field of biology; In addition, appropriate preparation of this salt is used as a radioactive isotope to study the perfusion of heart muscle. In the field of biochemistry, it is used to induce cells to take up DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 1390°C |
Melting Point | 718°C |
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Assay Percent Range | 99.975% (metals basis) |
Solubility Information | Soluble in water. |
Formula Weight | 120.92 |
Odor | Odorless |
Refractive Index | 1.493 |
Show More |
RUO – Research Use Only