missing translation for 'onlineSavingsMsg'
Learn More

Rubidium chloride, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

184.00 € - 653.00 €

Chemical Identifiers

CAS 7791-11-9
Molecular Formula ClRb
Molecular Weight (g/mol) 120.92
MDL Number MFCD00011187
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
PubChem CID 62683
ChEBI CHEBI:78672
IUPAC Name rubidium(1+) chloride
SMILES [Cl-].[Rb+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11366997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010851.09
10 g
184.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11376997
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010851.18
50 g
653.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rubidium chloride is the mostly used rubidium compound, and finds its applications in various fields ranging from electrochemistry to molecular biology. It is used as a non-invasive biomarker, anergic and apathetic depressives in the field of biology; In addition, appropriate preparation of this salt is used as a radioactive isotope to study the perfusion of heart muscle. In the field of biochemistry, it is used to induce cells to take up DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-11-9
120.92
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
62683
rubidium(1+) chloride
ClRb
MFCD00011187
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
CHEBI:78672
[Cl-].[Rb+]

Specifications

1390°C
7791-11-9
Crystalline Aggregates
ClRb
rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
rubidium(1+) chloride
62683
120.92
1.493
White crystals
Rubidium chloride
718°C
White
99.975% (metals basis)
MFCD00011187
Soluble in water.
[Cl-].[Rb+]
120.92
CHEBI:78672
Odorless
Hygroscopic
2.8 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 232-240-9

missing translation for 'rtecsNumber' : VL8575000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only