Learn More
Rubidium fluoride, 99.1% (metals basis), Thermo Scientific Chemicals
Description
It is used as raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 795°C |
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
Linear Formula | RbF |
UN Number | UN3288 |
Solubility Information | Soluble in water. (1306g/L at 20°C). |
Formula Weight | 104.47 |
Percent Purity | 99.1% |
Refractive Index | 1.398 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253527 | Certificate of Analysis | 25/12/2024 | 047144.09,047144.18 |
S06F047 | Certificate of Analysis | 25/11/2024 | 047144.09,047144.18 |
U31D036 | Certificate of Analysis | 08/12/2021 | 047144.09,047144.18 |
W30C023 | Certificate of Analysis | 07/12/2021 | 047144.09,047144.18 |
M05C016 | Certificate of Analysis | 07/12/2021 | 047144.09,047144.18 |
Safety and Handling

- Rubidium fluoride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Rubidium fluoride (RbF)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.