missing translation for 'onlineSavingsMsg'
Learn More

Rubidium hexafluoroarsenate(V), 98% (metals basis), Thermo Scientific™

Chemical Identifiers

CAS 43023-95-6
Molecular Formula AsF6Rb
Molecular Weight (g/mol) 274.38
MDL Number MFCD00042537
InChI Key FDUSNUSHTOTAKN-UHFFFAOYSA-N
Synonym rubidium hexafluoroarsenate, acmc-20ajsa, rubidium hexafluoroarsenate v, rubidium hexafluoroarsenate 1-, hexafluoroarsoranuide; rubidium 1+, arsenate 1-,hexafluoro-,rubidium 9ci, arsenate 1-,hexafluoro-, rubidium 9ci, rubidium 1+ hexafluoro-$l^ 5-arsanuide, hexakis fluoranyl arsenic 1-; rubidium 1+, rubidium 1+ ion hexafluoro-??-arsanuide
PubChem CID 14205728
IUPAC Name hexafluoroarsenic(1-);rubidium(1+)
SMILES F[As-](F)(F)(F)(F)F.[Rb+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398318
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039518.06
5 g N/A
N/A
11308328
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039518.14
25 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

43023-95-6
274.38
FDUSNUSHTOTAKN-UHFFFAOYSA-N
14205728
F[As-](F)(F)(F)(F)F.[Rb+]
AsF6Rb
MFCD00042537
rubidium hexafluoroarsenate, acmc-20ajsa, rubidium hexafluoroarsenate v, rubidium hexafluoroarsenate 1-, hexafluoroarsoranuide; rubidium 1+, arsenate 1-,hexafluoro-,rubidium 9ci, arsenate 1-,hexafluoro-, rubidium 9ci, rubidium 1+ hexafluoro-$l^ 5-arsanuide, hexakis fluoranyl arsenic 1-; rubidium 1+, rubidium 1+ ion hexafluoro-??-arsanuide
hexafluoroarsenic(1-);rubidium(1+)

Specifications

43023-95-6
AsF6Rb
UN1557
Soluble in water.
F[As-](F)(F)(F)(F)F.[Rb+]
274.38
359.92
(metals basis)
Powder
MFCD00042537
rubidium hexafluoroarsenate, acmc-20ajsa, rubidium hexafluoroarsenate v, rubidium hexafluoroarsenate 1-, hexafluoroarsoranuide; rubidium 1+, arsenate 1-,hexafluoro-,rubidium 9ci, arsenate 1-,hexafluoro-, rubidium 9ci, rubidium 1+ hexafluoro-$l^ 5-arsanuide, hexakis fluoranyl arsenic 1-; rubidium 1+, rubidium 1+ ion hexafluoro-??-arsanuide
FDUSNUSHTOTAKN-UHFFFAOYSA-N
hexafluoroarsenic(1-);rubidium(1+)
14205728
98%
Rubidium hexafluoroarsenate(V)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c

H301+H331

missing translation for 'dotInformation' : Hazard Class: 6.1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications