Learn More
Rubidium hydroxide hydrate, 99% (metals basis), Thermo Scientific Chemicals
Oxidative chlorination catalyst
Chemical Identifiers
CAS | 12026-05-0 |
---|---|
Molecular Formula | HORb |
Molecular Weight (g/mol) | 102.48 |
MDL Number | MFCD00011194 |
InChI Key | CPRMKOQKXYSDML-UHFFFAOYSA-M |
Synonym | rubidium hydroxide hydrate, rb.ho.h2o, rubidium hydroxide monohydrate, rubidium hydroxide-water 1/1/1, rubidium 1+ ion hydrate hydroxide, rubidium hydroxide rb oh , hydrate 9ci, rubidium hydroxide hydrate, fused solid-rb |
PubChem CID | 16212572 |
IUPAC Name | rubidium(1+);hydroxide;hydrate |
SMILES | [OH-].[Rb+] |
Description
Oxidative chlorination catalyst.Rubidium hydroxide hydrate is used in fireworks in place of pure rubidium to get violet color, thereby it prevents the waste of expensive element rubidium. It acts as an intermediate in the synthesis of other rubidium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12026-05-0 | |
102.48 | |
CPRMKOQKXYSDML-UHFFFAOYSA-M | |
16212572 | |
[OH-].[Rb+] |
HORb | |
MFCD00011194 | |
rubidium hydroxide hydrate, rb.ho.h2o, rubidium hydroxide monohydrate, rubidium hydroxide-water 1/1/1, rubidium 1+ ion hydrate hydroxide, rubidium hydroxide rb oh , hydrate 9ci, rubidium hydroxide hydrate, fused solid-rb | |
rubidium(1+);hydroxide;hydrate |
Specifications
12026-05-0 | |
Crystalline | |
RbOH·xH2O | |
UN2678 | |
rubidium hydroxide hydrate, rb.ho.h2o, rubidium hydroxide monohydrate, rubidium hydroxide-water 1/1/1, rubidium 1+ ion hydrate hydroxide, rubidium hydroxide rb oh , hydrate 9ci, rubidium hydroxide hydrate, fused solid-rb | |
CPRMKOQKXYSDML-UHFFFAOYSA-M | |
rubidium(1+);hydroxide;hydrate | |
16212572 | |
99% | |
(metals basis) |
White to Gray | |
HORb | |
MFCD00011194 | |
14,8285 | |
Soluble in water and alcohol. | |
[OH-].[Rb+] | |
102.48 | |
102.48 (Anhydrous) | |
Hygroscopic | |
Rubidium hydroxide hydrate |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE
missing translation for 'einecsNumber' : 215-186-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only