missing translation for 'onlineSavingsMsg'
Learn More
Rubidium nitrate, 99%, Thermo Scientific Chemicals Product Code.: 11397617

Rubidium nitrate, 99%, Thermo Scientific Chemicals

Product Code. 11397617
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11397617

Brand: Thermo Scientific Alfa Aesar 012889.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rubidium nitrate used in infrared radiation producing pyrotechnic compositions as a colorant and an oxidizer. It is also used as a raw material for preparation of other rubidium compounds and rubidium metal, for manufacture of catalysts and in scintillation counters. It is rarely used in fireworks to produce a red-violet colour.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13126-12-0
Molecular Formula NO3Rb
Molecular Weight (g/mol) 147.47
MDL Number MFCD00011193
InChI Key RTHYXYOJKHGZJT-UHFFFAOYSA-N
Synonym rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrateShow MoreShow Less
PubChem CID 25731
IUPAC Name rubidium(1+);nitrate
SMILES [Rb+].[O-][N+]([O-])=O

Specifications

Melting Point 310°C
Color White
Physical Form Crystalline
Quantity 250 g
UN Number UN1477
Solubility Information Soluble in water. Slightly soluble in acetone. Very soluble in nitric acid.
Formula Weight 147.47
Percent Purity 99.8%
Refractive Index 1.52
Sensitivity Hygroscopic
Density 3.11 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN16J001 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012889.30,012889.18,012889.09
Lot NumberB04N11 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012889.30,012889.18,012889.09
Lot NumberG12L31 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012889.30,012889.18,012889.09
Lot NumberT25D092 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012889.30,012889.18,012889.09
Lot NumberM19H003 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012889.30,012889.18,012889.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium nitrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.