Learn More
Rubidium nitrate, 99% (metals basis), Thermo Scientific Chemicals
As a catalyst in organic reactions and as chelating agents
95.30 € - 359.00 €
Chemical Identifiers
CAS | 13126-12-0 |
---|---|
Molecular Formula | NO3Rb |
Molecular Weight (g/mol) | 147.47 |
MDL Number | MFCD00011193 |
InChI Key | RTHYXYOJKHGZJT-UHFFFAOYSA-N |
Synonym | rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate |
PubChem CID | 25731 |
SMILES | [Rb+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346868
|
Thermo Scientific Alfa Aesar
013496.14 |
25 g |
95.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336868
|
Thermo Scientific Alfa Aesar
013496.22 |
100 g |
359.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rubidium nitrate is used in infrared radiation producing pyrotechnic compositions as a colorant and an oxidizer, e.g. in decoys and illumination flares. It is also used as a raw material for preparation of other rubidium compounds and rubidium metal, for manufacture of catalysts and in scintillation counters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13126-12-0 | |
147.47 | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
25731 |
NO3Rb | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
[Rb+].[O-][N+]([O-])=O |
Specifications
310°C | |
White | |
NO3Rb | |
UN1477 | |
Soluble in water; Slightly soluble in acetone; Very soluble in HNO3 | |
[Rb+].[O-][N+]([O-])=O | |
147.47 | |
147.47 | |
1.52 | |
3.11 g/mL | |
Rubidium nitrate |
13126-12-0 | |
Crystalline | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
rubidium(1+);nitrate | |
25731 | |
99% | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P220-P221-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335-H350
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 236-060-1
missing translation for 'rtecsNumber' : QV0900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only