Learn More
Rubidium nitrate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 13126-12-0 |
---|---|
Molecular Formula | NO3Rb |
Molecular Weight (g/mol) | 147.47 |
MDL Number | MFCD00011193 |
InChI Key | RTHYXYOJKHGZJT-UHFFFAOYSA-N |
Synonym | rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate |
PubChem CID | 25731 |
SMILES | [Rb+].[O-][N+]([O-])=O |
Description
Rubidium nitrate is used in infrared radiation producing pyrotechnic compositions. It acts as a colorant and an oxidizer especially in decoys and illumination flares. Further, it is employed as a starting material in the preparation of other rubidium compounds and rubidium metal, which is utilized for the preparation of catalysts and in scintillation counters. In addition to this, it is used in fireworks to produce a red-violet color.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13126-12-0 | |
147.47 | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
25731 |
NO3Rb | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
[Rb+].[O-][N+]([O-])=O |
Specifications
310°C | |
Crystalline | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
[Rb+].[O-][N+]([O-])=O | |
147.47 | |
147.47 | |
Puratronic™ | |
Hygroscopic | |
(metals basis) |
13126-12-0 | |
NO3Rb | |
UN1477 | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
rubidium(1+) nitrate | |
25731 | |
99.975% | |
1.52 | |
3.11g/mL | |
Rubidium nitrate |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
missing translation for 'rtecsNumber' : QV0900000
missing translation for 'tsca' : TSCA
RUO – Research Use Only