missing translation for 'onlineSavingsMsg'
Learn More

Rubidium sulfate, 99% (metals basis), Thermo Scientific Chemicals

99.70 € - 1381.88 €

Chemical Identifiers

CAS 7488-54-2
Molecular Formula H2O4RbS
Molecular Weight (g/mol) 183.54
MDL Number MFCD00011190
InChI Key XNNUAZGCOYTNQE-UHFFFAOYSA-N
PubChem CID 197088
IUPAC Name sulfuric acid rubidium
SMILES [Rb].OS(O)(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11316868
View Documents
Thermo Scientific Alfa Aesar
013493.14
25 g
99.70 €
25g
Please to purchase this item. Need a web account? Register with us today!
11326868
View Documents
Thermo Scientific Alfa Aesar
013493.22
100 g
331.65 €
100g
Please to purchase this item. Need a web account? Register with us today!
11450320
View Documents
Thermo Scientific Alfa Aesar
013493.36
500 g
1381.88 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7488-54-2
183.54
XNNUAZGCOYTNQE-UHFFFAOYSA-N
sulfuric acid rubidium
H2O4RbS
MFCD00011190
197088
[Rb].OS(O)(=O)=O

Specifications

1050°C
Crystalline
MFCD00011190
Soluble in water (424mg/ml at 10°C).
[Rb].OS(O)(=O)=O
183.54
267
Odorless
3.613 g/mL
Rubidium sulfate
7488-54-2
H2O4RbS
rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
XNNUAZGCOYTNQE-UHFFFAOYSA-N
sulfuric acid rubidium
197088
99%
1.513
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 231-301-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only