missing translation for 'onlineSavingsMsg'
Learn More

Rubidium tetrafluoroborate, 98%, Thermo Scientific Chemicals

76.88 € - 346.73 €

Chemical Identifiers

CAS 18909-68-7
Molecular Formula BF4Rb
Molecular Weight (g/mol) 172.27
MDL Number MFCD00083071
InChI Key MERZTPVDEIGCOQ-UHFFFAOYSA-N
Synonym rubidium tetrafluoroborate, rubidium 1+ tetrafluoroborate, rubidium fluoroborate, rubidium boron fluoride, borate 1-, rubidium, acmc-1bu19, rubidium 1+ ion tetrafluoroborate
PubChem CID 23696894
IUPAC Name rubidium(1+) tetrafluoroboranuide
SMILES [Rb+].F[B-](F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11445746
SDS View Documents
Thermo Scientific Alfa Aesar
B20817.06
5 g
76.88 €
5g
Please to purchase this item. Need a web account? Register with us today!
11455746
SDS View Documents
Thermo Scientific Alfa Aesar
B20817.14
25 g
346.73 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18909-68-7
172.27
MERZTPVDEIGCOQ-UHFFFAOYSA-N
23696894
[Rb+].F[B-](F)(F)F
BF4Rb
MFCD00083071
rubidium tetrafluoroborate, rubidium 1+ tetrafluoroborate, rubidium fluoroborate, rubidium boron fluoride, borate 1-, rubidium, acmc-1bu19, rubidium 1+ ion tetrafluoroborate
rubidium(1+) tetrafluoroboranuide

Specifications

18909-68-7
RbBF4
rubidium tetrafluoroborate, rubidium 1+ tetrafluoroborate, rubidium fluoroborate, rubidium boron fluoride, borate 1-, rubidium, acmc-1bu19, rubidium 1+ ion tetrafluoroborate
[Rb+].F[B-](F)(F)F
172.27
172.27
Rubidium tetrafluoroborate
BF4Rb
MFCD00083071
MERZTPVDEIGCOQ-UHFFFAOYSA-N
rubidium(1+) tetrafluoroboranuide
23696894
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 242-660-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.