missing translation for 'onlineSavingsMsg'
Learn More

uvex Rubiflex S NB60S Chemical Protection Gloves

Product Code. 17250242 Shop All Uvex Products
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Long, NBR-coated chemical protection gloves feature reinforced cotton interlock supporting material for applications that require protection against chemicals and mechanical hazards.

  • NBR-coated chemical protection gloves with an extended gauntlet cuff
  • Reinforced cotton interlock supporting material provides mechanical protection
  • Resistant to many chemicals, acids, alkalis, mineral oils, and solvents
  • Ergonomic design is comfortable and highly flexible
  • Silicone-free according to imprint test
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 8
EN388 Standard 2121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKOPT
For Use With (Application) Chemical Handling, Municipal Cleaning, Sandblasting, and Sewer Construction
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex S NB60S
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.