Learn More
Brady™ Rugged Reinforced Nylon Safety Padlocks
Brady™ Standard Safety Padlocks with Steel Shackle are the ultimate locks for all safety and security needs, and are precision-machined for better tamper resistance.
Brand:  Brady™ 813596
 View more versions of this product
Product Code. 15613557
  • 145.00 € / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique cylinder is insulated to protect workers from shocks when key is inserted
  • Six-pin precision-machined for more unique key cuts and better tamper resistance
  • Reserved, paracentric keyway provides optimal security
  • Special design prohibits the key from being released until the padlock shackle is closed
  • Locks are compact and lightweight and resist impact, temperature, chemicals and corrosion
  • Non-conductive, non-sparking body features a ribbed design for easier gripping and handling
  • One key supplied with each lock
  • Key retaining - key will not release until the padlock shackle is securely closed
Specifications
Specifications
Safety Padlocks
Yellow
Safety Padlocks - Nylon Shackle
38.00 x 86.50 mm
38.00 mm
6 Pk.
33 mm
SFTY PADLOCK NYLON 38MM SHA YELLOW 6/PAC
Reinforced Nylon
6.5 mm
86.5 mm
6
38 mm