missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Rulers

Moulded in white plastic.

Brand:  Fisherbrand™ Z7904/8304

 View more versions of this product

Product Code. 12989074

  • 74.45 € / Pack of 10
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • 6mm thick by 25mm wide
  • 1 meter long
  • Foil printed in black

Specifications

Specifications

Ruler
1 m
6 mm
Plastic
White
Metric
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers