missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, 0.5% on granular carbon, Thermo Scientific Chemicals

228.14 € - 634.16 €

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314457
View Documents
Thermo Scientific Alfa Aesar
038332.14
25 g
228.14 €
25g
Please to purchase this item. Need a web account? Register with us today!
11304457
View Documents
Thermo Scientific Alfa Aesar
038332.22
100 g
634.16 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ruthenium is used in wear-resistant electrical contacts and the production of thick-film resistors. Its use in some platinum alloys, and as a catalyst. It is a most effective hardeners for platinum and palladium. It is also used in some advanced high-temperature single-crystal super alloys, with applications including the turbine blades in jet engines and fountain pen nibs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]

Specifications

7440-18-8
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
0.5% on granular carbon
Ruthenium
Ru
UN1325
Soluble in fused alkalis. Insoluble in water,acid and aqua regia.
[Ru]
101.07
CHEBI:30682
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-127-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only