missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) 2,4-pentanedionate, Ru 24% min, Thermo Scientific Chemicals

For Hydrogenation

130.65 € - 577.88 €

Chemical Identifiers

CAS 14284-93-6
Molecular Formula C15H21O6Ru
Molecular Weight (g/mol) 398.40
MDL Number MFCD00000030
InChI Key RTZYCRSRNSTRGC-LNTINUHCSA-K
Synonym ruthenium acetylacetonate, ruthenium iii acetylacetonate, acetylacetone ruthenium iii salt, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-8ci, z-4-oxopent-2-en-2-olate; ruthenium 3+
PubChem CID 5488829
SMILES [Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11300729
SDS View Documents
Thermo Scientific Alfa Aesar
010568.03
1 g
130.65 €
1g
Please to purchase this item. Need a web account? Register with us today!
11310729
SDS View Documents
Thermo Scientific Alfa Aesar
010568.06
5 g
577.88 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Precursor for the preparation of other compounds of ruthenium. Ruthenium(III) 2,4-pentanedionate is employed as a recyclable catalyst for certain organic transformations like the acetylation of phenols, alcohols, and amines under neat conditions. It is also used as a homogeneous catalyst, for example, in the hydrolysis of sodium borohydride and regiospecific tritiation of aromatic carboxylic acids. It is also used as a catalyst for enantioselective hydrogenation of acids such as aryl acrylic acid and aryl propenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14284-93-6
398.40
RTZYCRSRNSTRGC-LNTINUHCSA-K
5488829
C15H21O6Ru
MFCD00000030
ruthenium acetylacetonate, ruthenium iii acetylacetonate, acetylacetone ruthenium iii salt, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-8ci, z-4-oxopent-2-en-2-olate; ruthenium 3+
[Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

260°C (decomposition)
White
C15H21O6Ru
ruthenium acetylacetonate, ruthenium iii acetylacetonate, acetylacetone ruthenium iii salt, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-8ci, z-4-oxopent-2-en-2-olate; ruthenium 3+
RTZYCRSRNSTRGC-LNTINUHCSA-K
(Z)-4-oxopent-2-en-2-olate;ruthenium(3+)
5488829
Red crystals
14284-93-6
Crystalline
MFCD00000030
Soluble in most organic solvents such as acetone,chlorinated hydrocarbons,alcohols,cyclohexane and benzene. Insoluble in water.
[Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
398.40
398.4
Ruthenium(III) 2, 4-pentanedionate, Ru 24% min.
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 238-193-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.